missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)piperidine 97.0+%, TCI America™

Catalog No. T30811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30811G 1 g
T30815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30811G Supplier TCI America Supplier No. T30811G
Only null left

Chemical Identifiers

CAS 657-36-3
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
MDL Number MFCD00102068
InChI Key RDRQUUWCJTYHCT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
PubChem CID 136469
IUPAC Name 4-(trifluoromethyl)piperidine
SMILES FC(F)(F)C1CCNCC1

Specifications

Color Yellow
Boiling Point 135°C
Quantity 1 g
UN Number 1993
Formula Weight 153.15
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)piperidine