missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)piperidine, 97%

Catalog No. p-7056089
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64840MD 250 mg
AAH6484003 1 g
AAH6484006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64840MD Supplier Thermo Scientific Chemicals Supplier No. H64840MD

CAS: 657-36-3 | C6H10F3N | 153.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 657-36-3
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
MDL Number MFCD00102068
InChI Key RDRQUUWCJTYHCT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
PubChem CID 136469
IUPAC Name 4-(trifluoromethyl)piperidine
SMILES FC(F)(F)C1CCNCC1

Specifications

Color Colorless
Quantity 250 mg
Formula Weight 153.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only