missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylhydrazine hydrochloride, 96%

Catalog No. p-7031309
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3234003 1 g
AAH3234009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3234003 Supplier Thermo Scientific Chemicals Supplier No. H3234003

CAS: 2923-56-0 | C7H8ClF3N2 | 212.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2923-56-0
Molecular Formula C7H8ClF3N2
Molecular Weight (g/mol) 212.6
MDL Number MFCD00204233
InChI Key WCAGNYIHAYOPSE-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylhydrazine hydrochloride, 4-trifluoromethyl phenyl hydrazine hydrochloride, 1-4-trifluoromethyl phenyl hydrazine hydrochloride, 4-trifluoromethylphenyl hydrazine hydrochloride, 4-trifluoromethyl phenylhydrazine hcl, 4-trifluoromethyl phenylhydrazine, hcl, hydrazine, 4-trifluoromethyl phenyl-, monohydrochloride, n'-4-trifluoromethyl-phenyl-hydrazinium, chloride
PubChem CID 24229790
IUPAC Name [4-(trifluoromethyl)phenyl]hydrazine;hydrochloride
SMILES C1=CC(=CC=C1C(F)(F)F)NN.Cl

Specifications

Melting Point ∽210°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 212.6
Percent Purity 96%
Chemical Name or Material 4-(Trifluoromethyl)phenylhydrazine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 642-499-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only