missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylhydrazine, 95%

Catalog No. AAL1243603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1243603 1 g
AAL1243606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1243603 Supplier Thermo Scientific Chemicals Supplier No. L1243603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 368-90-1 | C7H7F3N2 | 176.14 g/mol

4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 368-90-1
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.14
MDL Number MFCD00042508
InChI Key DBNLGTYGKCMLLR-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylhydrazine, 4-trifluoromethyl phenyl hydrazine, 4-trifluoromethylphenylhydrazine, p-trifluoromethylphenylhydrazine, hydrazine, 4-trifluoromethyl phenyl, p-trifluoromethyl phenylhydrazine, 4-trifluoromethyl-phenylhydrazine, 4-trifluoromethyl-phenyl-hydrazine
PubChem CID 136200
IUPAC Name [4-(trifluoromethyl)phenyl]hydrazine
SMILES NNC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 60°C to 64°C
Boiling Point 119°C to 121°C (17 mmHg)
Quantity 1 g
Beilstein 743759
Solubility Information Insoluble in water.
Formula Weight 176.14
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethyl)phenylhydrazine
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only