missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis

Catalog No. AAL2027506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2027506 5 g
AAL2027503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2027506 Supplier Thermo Scientific Chemicals Supplier No. L2027506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1736-56-7 | C9H5F3O2 | 202.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1736-56-7
Molecular Formula C9H5F3O2
Molecular Weight (g/mol) 202.132
MDL Number MFCD05664098
InChI Key BGOMXTCPIUNFKR-UHFFFAOYSA-N
Synonym 2-oxo-2-4-trifluoromethyl phenyl acetaldehyde, 2-oxo-2-4-trifluoromethyl phenyl ethanal, 2-oxidanylidene-2-4-trifluoromethyl phenyl ethanal, 4-trifluoromethylphenyl oxo acetaldehyde, oxo 4-trifluoromethyl phenyl acetaldehyde, 1-4-trifluoromethyl phenyl-1,2-ethanedione
PubChem CID 2783286
IUPAC Name 2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde
SMILES C1=CC(=CC=C1C(=O)C=O)C(F)(F)F

Specifications

Melting Point 83°C to 85°C
Quantity 5 g
Beilstein 2261131
Formula Weight 202.13 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 4-(Trifluoromethyl)phenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only