missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Melting Point | 250°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | 4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid |
| SMILES | OB(O)C1=CC=C(C=C1)C(F)(F)F |
| Molecular Weight (g/mol) | 189.93 |
| Formula Weight | 189.93 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | Yes |