missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T17881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17881G 1 g
T17885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17881G Supplier TCI America Supplier No. T17881G

Chemical Identifiers

CAS 128796-39-4
Molecular Formula C7H6BF3O2
MDL Number MFCD00151855
InChI Key ALMFIOZYDASRRC-UHFFFAOYSA-N
PubChem CID 2734389
IUPAC Name [4-(trifluoromethyl)phenyl]boronic acid

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 1 g
Synonym 4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 189.93
Formula Weight 189.93
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)