missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylacetylene, 97%

Catalog No. AC435370050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435370050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435370050 Supplier Thermo Scientific Chemicals Supplier No. 435370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 705-31-7 | C9H5F3 | 170.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 705-31-7
Molecular Formula C9H5F3
Molecular Weight (g/mol) 170.13
MDL Number MFCD01861903
InChI Key XTKBMZQCDBHHKY-UHFFFAOYSA-N
Synonym 1-ethynyl-4-trifluoromethyl benzene, 4'-trifluoromethylphenyl acetylene, 4-trifluoromethyl phenylacetylene, 4-ethynyl-alpha,alpha,alpha-trifluorotoluene, 4-ethynylbenzotrifluoride, 4-ethynyl-a,a,a-trifluorotoluene, 4-ethynyl trifluorotoluene, benzene, 1-ethynyl-4-trifluoromethyl, 1-ethynyl-4-trifluoromethyl-benzene, 4-ethynyl-1-trifluoromethyl benzene
PubChem CID 4348295
IUPAC Name 1-ethynyl-4-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 58.0°C to 60.0°C (30.0 Torr)
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 170.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)phenylacetylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only