missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylacetic Acid 97.0+%, TCI America™

Catalog No. T17831G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17831G 1 g
T17835G 5 g
T178325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T17831G Supplier TCI America Supplier No. T17831G

Chemical Identifiers

CAS 32857-62-8
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00004352
InChI Key HNORVZDAANCHAY-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylacetic acid, 2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylphenylacetic acid, 4-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-p-tolyl acetic acid, benzeneacetic acid, 4-trifluoromethyl, p-trifluoromethyl phenylacetic acid, p-trifluoromethylphenylacetic acid, alpha,alpha,alpha-trifluoro-4-tolyl acetic acid
PubChem CID 118342
IUPAC Name 2-[4-(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 83°C
Quantity 1 g
Formula Weight 204.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)phenylacetic Acid