missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7137226
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24611G 1 g
T24615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24611G Supplier TCI America Supplier No. T24611G
Only null left

Chemical Identifiers

CAS 1645-65-4
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
MDL Number MFCD00039645
InChI Key DQEVDFQAYLIBRD-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethyl benzene, 4-trifluoromethyl phenylisothiocyanate, 4-trifluoromethylphenylisothiocyanate, p-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolyl isothiocyanate, isothiocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethyl-benzene
PubChem CID 137134
IUPAC Name 1-isothiocyanato-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)N=C=S

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 207°C
Quantity 1 g
UN Number 2811
Formula Weight 203.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)phenyl Isothiocyanate
TSCA No
Recommended Storage Refrigerator