missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™
$236.73 - $700.89
Chemical Identifiers
| CAS | 1232133-62-8 |
|---|---|
| Molecular Formula | C17H15F6IO3S |
| Molecular Weight (g/mol) | 540.259 |
| MDL Number | MFCD27978254 |
| InChI Key | XTTMIKRDUJQRGJ-UHFFFAOYSA-M |
| Synonym | Mesityl[4-(trifluoromethyl)phenyl]iodonium Triflate |
| PubChem CID | 117064601 |
| IUPAC Name | trifluoromethanesulfonate;[4-(trifluoromethyl)phenyl]-(2,4,6-trimethylphenyl)iodanium |
| SMILES | CC1=CC(=C(C(=C1)C)[I+]C2=CC=C(C=C2)C(F)(F)F)C.C(F)(F)(F)S(=O)(=O)[O-] |
Chemical Identifiers
| 1232133-62-8 | |
| 540.259 | |
| XTTMIKRDUJQRGJ-UHFFFAOYSA-M | |
| 117064601 | |
| CC1=CC(=C(C(=C1)C)[I+]C2=CC=C(C=C2)C(F)(F)F)C.C(F)(F)(F)S(=O)(=O)[O-] |
| C17H15F6IO3S | |
| MFCD27978254 | |
| Mesityl[4-(trifluoromethyl)phenyl]iodonium Triflate | |
| trifluoromethanesulfonate;[4-(trifluoromethyl)phenyl]-(2,4,6-trimethylphenyl)iodanium |
Specifications
| 1232133-62-8 | |
| C17H15F6IO3S | |
| 1 g | |
| XTTMIKRDUJQRGJ-UHFFFAOYSA-M | |
| trifluoromethanesulfonate;[4-(trifluoromethyl)phenyl]-(2,4,6-trimethylphenyl)iodanium | |
| 117064601 | |
| ≥98.0% (T) | |
| [4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate |
| 203°C | |
| MFCD27978254 | |
| Mesityl[4-(trifluoromethyl)phenyl]iodonium Triflate | |
| CC1=CC(=C(C(=C1)C)[I+]C2=CC=C(C=C2)C(F)(F)F)C.C(F)(F)(F)S(=O)(=O)[O-] | |
| 540.259 | |
| 540.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No