CAS RN 1232133-62-8
CAS RN 1232133-62-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™
CAS: 1232133-62-8 Molecular Formula: C17H15F6IO3S Molecular Weight (g/mol): 540.259 MDL Number: MFCD27978254 InChI Key: XTTMIKRDUJQRGJ-UHFFFAOYSA-M Synonym: Mesityl[4-(trifluoromethyl)phenyl]iodonium Triflate PubChem CID: 117064601 IUPAC Name: trifluoromethanesulfonate;[4-(trifluoromethyl)phenyl]-(2,4,6-trimethylphenyl)iodanium SMILES: CC1=CC(=C(C(=C1)C)[I+]C2=CC=C(C=C2)C(F)(F)F)C.C(F)(F)(F)S(=O)(=O)[O-]