missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenol, 98%

CAS: 402-45-9 | C7H5F3O | 162.11 g/mol

Supplier:  Thermo Scientific Chemicals 176440050

 View more versions of this product

Catalog No. AC176440050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Trifluoromethyl)phenol
97.5
40.0°C to 46.0°C
71.5°C to 72.0°C (8.0 mmHg)
Authentic
Glass bottle
CF3C6H4OH
5 g
BAYGVMXZJBFEMB-UHFFFAOYSA-N
4-(trifluoromethyl)phenol
67874
162.11
Crystals
402-45-9
100.0
83g/mL
83°C
97.5% min. (GC)
C7H5F3O
MFCD00002363
4-trifluoromethyl phenol, 4-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-p-cresol, p-trifluoromethylphenol, p-hydroxybenzotrifluoride, phenol, 4-trifluoromethyl, a,a,a-trifluoro-p-cresol, 4-trifluoromethyl-phenol, para-trifluoromethylphenol
C1=CC(=CC=C1C(F)(F)F)O
162.11
CHEBI:42578
98%

Chemical Identifiers

402-45-9
162.11
BAYGVMXZJBFEMB-UHFFFAOYSA-N
67874
4-(trifluoromethyl)phenol
C7H5F3O
MFCD00002363
4-trifluoromethyl phenol, 4-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-p-cresol, p-trifluoromethylphenol, p-hydroxybenzotrifluoride, phenol, 4-trifluoromethyl, a,a,a-trifluoro-p-cresol, 4-trifluoromethyl-phenol, para-trifluoromethylphenol
CHEBI:42578
C1=CC(=CC=C1C(F)(F)F)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye damage.
Toxic if swallowed.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash wi

GHS Signal Word: Danger

EINECSNumber : 206-945-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only