missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenol, 98%

Catalog No. AC176440010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176440010 1 g Glass bottle
AC176440050 5 g Glass bottle
AC176440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC176440010 Supplier Thermo Scientific Chemicals Supplier No. 176440010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 402-45-9 | C7H5F3O | 162.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-45-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.11
MDL Number MFCD00002363
InChI Key BAYGVMXZJBFEMB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenol, 4-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-p-cresol, p-trifluoromethylphenol, p-hydroxybenzotrifluoride, phenol, 4-trifluoromethyl, a,a,a-trifluoro-p-cresol, 4-trifluoromethyl-phenol, para-trifluoromethylphenol
PubChem CID 67874
ChEBI CHEBI:42578
IUPAC Name 4-(trifluoromethyl)phenol
SMILES C1=CC(=CC=C1C(F)(F)F)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 40.0°C to 46.0°C
Density 83g/mL
Boiling Point 71.5°C to 72.0°C (8.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4OH
Quantity 1 g
Formula Weight 162.11
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 4-(Trifluoromethyl)phenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash wi
EINECSNumber 206-945-7

RUO – Research Use Only