missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)cyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. T36131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36131G 1 g
T36135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36131G Supplier TCI America Supplier No. T36131G

Chemical Identifiers

CAS 58665-70-6
Molecular Formula C7H12F3N
Molecular Weight (g/mol) 167.175
MDL Number MFCD09040573
InChI Key YCBWLMWEQURJHX-UHFFFAOYSA-N
PubChem CID 13714353
IUPAC Name 4-(trifluoromethyl)cyclohexan-1-amine
SMILES C1CC(CCC1C(F)(F)F)N

Specifications

Color Yellow
Boiling Point 71°C
Quantity 1 g
UN Number 2734
Formula Weight 167.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)cyclohexylamine (cis- and trans- mixture)