missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. T27065G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27065G Supplier TCI America Supplier No. T27065G

Chemical Identifiers

CAS 30129-18-1
Molecular Formula C7H11F3O
Molecular Weight (g/mol) 168.159
MDL Number MFCD00102144
InChI Key VJUJYNJEPPWWHS-UHFFFAOYSA-N
Synonym 4-trifluoromethyl cyclohexanol, trans-4-trifluoromethyl cyclohexanol, 4-trifluoromethyl cyclohexan-1-ol, 4-trifluoromethylcyclohexanol, 1r,4r-4-trifluoromethyl cyclohexanol, 4-trifluoromethyl-cyclohexanol, 4-trifluoromethyl cyclohexanol, cis/trans, cyclohexanol, 4-trifluoromethyl, cyclohexanol, 4-trifluoromethyl-, cis, cyclohexanol,4-trifluoromethyl-, trans
PubChem CID 2779765
IUPAC Name 4-(trifluoromethyl)cyclohexan-1-ol
SMILES C1CC(CCC1C(F)(F)F)O

Specifications

Color Colorless
Boiling Point 181°C
Quantity 5 g
Formula Weight 168.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)