accessibility menu, dialog, popup

UserName

FFFOH

CAS RN 30129-18-1

IUPAC Name: 4-(trifluoromethyl)cyclohexan-1-ol
Synonyms: 4-(trifluoromethyl)cyclohexan-1-ol
Molecular Weight (g/mol): 168.16
Molecular Formula: C7H11F3O
InChi Key: VJUJYNJEPPWWHS-UHFFFAOYSA-N
SMILES: OC1CCC(CC1)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)