missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)cyclohexanol, 97%

Catalog No. p-7032006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3331703 1 g
AAH3331706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3331703 Supplier Thermo Scientific Chemicals Supplier No. H3331703
Only null left

CAS: 30129-18-1 | C7H11F3O | 168.159 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 30129-18-1
Molecular Formula C7H11F3O
Molecular Weight (g/mol) 168.159
MDL Number MFCD00102144
InChI Key VJUJYNJEPPWWHS-UHFFFAOYSA-N
Synonym 4-trifluoromethyl cyclohexanol, trans-4-trifluoromethyl cyclohexanol, 4-trifluoromethyl cyclohexan-1-ol, 4-trifluoromethylcyclohexanol, 1r,4r-4-trifluoromethyl cyclohexanol, 4-trifluoromethyl-cyclohexanol, 4-trifluoromethyl cyclohexanol, cis/trans, cyclohexanol, 4-trifluoromethyl, cyclohexanol, 4-trifluoromethyl-, cis, cyclohexanol,4-trifluoromethyl-, trans
PubChem CID 2779765
IUPAC Name 4-(trifluoromethyl)cyclohexan-1-ol
SMILES C1CC(CCC1C(F)(F)F)O

Specifications

Boiling Point 70°C to 75°C (15 mmHg)
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 168.16
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)cyclohexanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only