missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)biphenyl-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. T32311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32311G 1 g
T32315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32311G Supplier TCI America Supplier No. T32311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84392-17-6
Molecular Formula C14H9F3O2
Molecular Weight (g/mol) 266.22
MDL Number MFCD00075353
InChI Key IQOMYCGTGFGDFN-UHFFFAOYSA-N
Synonym xenalipin, 4-trifluoromethyl-2'-biphenylcarboxylic acid, xenalipine, 4'-trifluoromethyl-1,1'-biphenyl-2-carboxylic acid, 2-4-trifluoromethyl phenyl benzoic acid, 4'-trifluoromethyl biphenyl-2-carboxylic acid, 4'-trifluoromethyl-2-biphenylcarboxylic acid, 2-4-trifluoromethylphenyl benzoic acid, 4'-trifluoromethyl 1,1'-biphenyl-2-carboxylic acid, 1,1'-biphenyl-2-carboxylic acid, 4'-trifluoromethyl
PubChem CID 55251
IUPAC Name 4'-(trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid
SMILES OC(=O)C1=CC=CC=C1C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 171°C
Color White
Quantity 1 g
Formula Weight 266.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(Trifluoromethyl)biphenyl-2-carboxylic Acid