missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzyl Bromide 98.0+%, TCI America™
$96.26 - $348.46
Chemical Identifiers
| CAS | 402-49-3 |
|---|---|
| Molecular Formula | C8H6BrF3 |
| Molecular Weight (g/mol) | 239.035 |
| MDL Number | MFCD00000403 |
| InChI Key | IKSNDOVDVVPSMA-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene |
| PubChem CID | 123062 |
| IUPAC Name | 1-(bromomethyl)-4-(trifluoromethyl)benzene |
| SMILES | C1=CC(=CC=C1CBr)C(F)(F)F |
Chemical Identifiers
| 402-49-3 | |
| 239.035 | |
| IKSNDOVDVVPSMA-UHFFFAOYSA-N | |
| 123062 | |
| C1=CC(=CC=C1CBr)C(F)(F)F |
| C8H6BrF3 | |
| MFCD00000403 | |
| 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene | |
| 1-(bromomethyl)-4-(trifluoromethyl)benzene |
Specifications
| 402-49-3 | |
| 66°C | |
| MFCD00000403 | |
| 3261 | |
| IKSNDOVDVVPSMA-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-(trifluoromethyl)benzene | |
| 123062 | |
| ≥98.0% (GC) | |
| 4-(Trifluoromethyl)benzyl Bromide |
| White-Yellow | |
| C8H6BrF3 | |
| 5 g | |
| 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene | |
| C1=CC(=CC=C1CBr)C(F)(F)F | |
| 239.035 | |
| 239.04 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No