Learn More
4-(Trifluoromethyl)benzyl bromide, 98%
CAS: 402-49-3 | C8H6BrF3 | 239.035 g/mol
$139.29 - $448.38
Chemical Identifiers
| CAS | 402-49-3 |
|---|---|
| Molecular Formula | C8H6BrF3 |
| Molecular Weight (g/mol) | 239.035 |
| MDL Number | MFCD00000403 |
| InChI Key | IKSNDOVDVVPSMA-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene |
| PubChem CID | 123062 |
| IUPAC Name | 1-(bromomethyl)-4-(trifluoromethyl)benzene |
| SMILES | C1=CC(=CC=C1CBr)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1689506
|
Thermo Scientific Chemicals
A1689506 |
5 g |
Each for $139.29
|
|
|||||
|
AAA1689514
|
Thermo Scientific Chemicals
A1689514 |
25 g |
Each for $448.38
|
|
|||||
Description
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents. Incompatible with oxidizing agents and bases.
Chemical Identifiers
| 402-49-3 | |
| 239.035 | |
| IKSNDOVDVVPSMA-UHFFFAOYSA-N | |
| 123062 | |
| C1=CC(=CC=C1CBr)C(F)(F)F |
| C8H6BrF3 | |
| MFCD00000403 | |
| 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene | |
| 1-(bromomethyl)-4-(trifluoromethyl)benzene |
Specifications
| 402-49-3 | |
| 1.546 | |
| 88°C (190°F) | |
| C8H6BrF3 | |
| MFCD00000403 | |
| UN3261 | |
| 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene | |
| IKSNDOVDVVPSMA-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-(trifluoromethyl)benzene | |
| 123062 | |
| 98% |
| 29°C to 33°C | |
| 65°C to 66°C (5 mmHg) | |
| Pungent | |
| 1.4915 | |
| 5 g | |
| 638980 | |
| Insoluble in water. | |
| C1=CC(=CC=C1CBr)C(F)(F)F | |
| 239.035 | |
| 239.04 | |
| 4-(Trifluoromethyl)benzyl bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-947-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only