missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl bromide, 98%

Catalog No. AAA1689506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1689506 5 g
AAA1689514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1689506 Supplier Thermo Scientific Chemicals Supplier No. A1689506

CAS: 402-49-3 | C8H6BrF3 | 239.035 g/mol

4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 402-49-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.035
MDL Number MFCD00000403
InChI Key IKSNDOVDVVPSMA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
PubChem CID 123062
IUPAC Name 1-(bromomethyl)-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1CBr)C(F)(F)F

Specifications

Melting Point 29°C to 33°C
Density 1.546
Boiling Point 65°C to 66°C (5 mmHg)
Flash Point 88°C (190°F)
Odor Pungent
Refractive Index 1.4915
Quantity 5 g
UN Number UN3261
Beilstein 638980
Solubility Information Insoluble in water.
Formula Weight 239.04
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzyl bromide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-947-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only