missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoyl chloride, 97%

Catalog No. AC303420010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303420010 1 g Glass bottle
AC303420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303420010 Supplier Thermo Scientific Chemicals Supplier No. 303420010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 329-15-7
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD00000694
InChI Key OXZYBOLWRXENKT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride
PubChem CID 67607
IUPAC Name 4-(trifluoromethyl)benzoyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)C(Cl)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 78°C to 79°C (16.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4755 to 1.4775
Quantity 1 g
Specific Gravity 1.4
Solubility Information Solubility in water: decomposes
Formula Weight 208.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)benzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-342-9

RUO – Research Use Only