Learn More
4-(Trifluoromethyl)benzoyl chloride, 97%
CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol
$115.99 - $381.30
Chemical Identifiers
| CAS | 329-15-7 |
|---|---|
| Molecular Formula | C8H4ClF3O |
| Molecular Weight (g/mol) | 208.56 |
| MDL Number | MFCD00000694 |
| InChI Key | OXZYBOLWRXENKT-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride |
| PubChem CID | 67607 |
| IUPAC Name | 4-(trifluoromethyl)benzoyl chloride |
| SMILES | FC(F)(F)C1=CC=C(C=C1)C(Cl)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1417606
|
Thermo Scientific Chemicals
A1417606 |
5 g |
Each for $115.99
|
|
|||||
|
AAA1417614
|
Thermo Scientific Chemicals
A1417614 |
25 g |
Each for $381.30
|
|
|||||
Description
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store under dry inert gas. Protect from humidity and water. Incompatible with water, bases, alcohol and oxidizing agents.
Chemical Identifiers
| 329-15-7 | |
| 208.56 | |
| OXZYBOLWRXENKT-UHFFFAOYSA-N | |
| 67607 | |
| FC(F)(F)C1=CC=C(C=C1)C(Cl)=O |
| C8H4ClF3O | |
| MFCD00000694 | |
| 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride | |
| 4-(trifluoromethyl)benzoyl chloride |
Specifications
| 329-15-7 | |
| 1.404 | |
| 78°C (172°F) | |
| C8H4ClF3O | |
| MFCD00000694 | |
| UN3265 | |
| Moisture sensitive | |
| Reacts with water. | |
| FC(F)(F)C1=CC=C(C=C1)C(Cl)=O | |
| 208.56 | |
| 208.57 | |
| 4-(Trifluoromethyl)benzoyl chloride |
| -3°C | |
| 188°C to 190°C | |
| Pungent | |
| 1.477 | |
| 5 g | |
| 391282 | |
| 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride | |
| OXZYBOLWRXENKT-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzoyl chloride | |
| 67607 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-342-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only