missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzonitrile 98.0+%, TCI America™
$51.98 - $86.64
Chemical Identifiers
| CAS | 455-18-5 |
|---|---|
| Molecular Formula | C8H4F3N |
| Molecular Weight (g/mol) | 171.122 |
| MDL Number | MFCD00001826 |
| InChI Key | DRNJIKRLQJRKMM-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzonitrile, benzonitrile, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolunitrile, 4-cyanobenzotrifluoride, p-trifluoromethylbenzonitrile, p-trifluoromethyl benzonitrile, benzonitrile, p-trifluoromethyl, p-trifluoromethylbenonitrile, para-trifluoromethyl-benzonitrile, p-cyanobenzotrifluoride |
| PubChem CID | 67995 |
| IUPAC Name | 4-(trifluoromethyl)benzonitrile |
| SMILES | C1=CC(=CC=C1C#N)C(F)(F)F |
Chemical Identifiers
| 455-18-5 | |
| 171.122 | |
| DRNJIKRLQJRKMM-UHFFFAOYSA-N | |
| 67995 | |
| C1=CC(=CC=C1C#N)C(F)(F)F |
| C8H4F3N | |
| MFCD00001826 | |
| 4-trifluoromethyl benzonitrile, benzonitrile, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolunitrile, 4-cyanobenzotrifluoride, p-trifluoromethylbenzonitrile, p-trifluoromethyl benzonitrile, benzonitrile, p-trifluoromethyl, p-trifluoromethylbenonitrile, para-trifluoromethyl-benzonitrile, p-cyanobenzotrifluoride | |
| 4-(trifluoromethyl)benzonitrile |
Specifications
| 455-18-5 | |
| 81°C | |
| MFCD00001826 | |
| 3439 | |
| DRNJIKRLQJRKMM-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzonitrile | |
| 67995 | |
| ≥98.0% (GC) | |
| 4-(Trifluoromethyl)benzonitrile |
| White-Yellow | |
| C8H4F3N | |
| 5 g | |
| 4-trifluoromethyl benzonitrile, benzonitrile, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolunitrile, 4-cyanobenzotrifluoride, p-trifluoromethylbenzonitrile, p-trifluoromethyl benzonitrile, benzonitrile, p-trifluoromethyl, p-trifluoromethylbenonitrile, para-trifluoromethyl-benzonitrile, p-cyanobenzotrifluoride | |
| C1=CC(=CC=C1C#N)C(F)(F)F | |
| 171.122 | |
| 171.12 | |
| Crystalline Lumps |
Safety and Handling
TSCA : Yes