missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. C10055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10055G 5 g
C100525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10055G Supplier TCI America Supplier No. C10055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 455-18-5
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001826
InChI Key DRNJIKRLQJRKMM-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzonitrile, benzonitrile, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolunitrile, 4-cyanobenzotrifluoride, p-trifluoromethylbenzonitrile, p-trifluoromethyl benzonitrile, benzonitrile, p-trifluoromethyl, p-trifluoromethylbenonitrile, para-trifluoromethyl-benzonitrile, p-cyanobenzotrifluoride
PubChem CID 67995
IUPAC Name 4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C(F)(F)F

Specifications

Color White-Yellow
Boiling Point 81°C
Quantity 5 g
UN Number 3439
Formula Weight 171.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(Trifluoromethyl)benzonitrile