missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzonitrile, 98%

Catalog No. AAA1451409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1451409 10 g
AAA1451418 50 g
AAA1451430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1451409 Supplier Thermo Scientific Chemicals Supplier No. A1451409

CAS: 455-18-5 | C8H4F3N | 171.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 455-18-5
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001826
InChI Key DRNJIKRLQJRKMM-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzonitrile, benzonitrile, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-tolunitrile, 4-cyanobenzotrifluoride, p-trifluoromethylbenzonitrile, p-trifluoromethyl benzonitrile, benzonitrile, p-trifluoromethyl, p-trifluoromethylbenonitrile, para-trifluoromethyl-benzonitrile, p-cyanobenzotrifluoride
PubChem CID 67995
IUPAC Name 4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C(F)(F)F

Specifications

Melting Point 36°C to 40°C
Density 1.278
Boiling Point 183°C to 185°C
Flash Point 71°C (159°F)
Refractive Index 1.4583
Quantity 10 g
UN Number UN3439
Beilstein 2046478
Formula Weight 171.12
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 207-239-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only