missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. T11455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11455G 5 g
T114525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11455G Supplier TCI America Supplier No. T11455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 455-24-3
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00002562
InChI Key SWKPKONEIZGROQ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride
PubChem CID 9966
ChEBI CHEBI:60696
IUPAC Name 4-(trifluoromethyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)C(F)(F)F

Specifications

Melting Point 223°C
Color White
Quantity 5 g
Formula Weight 190.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)benzoic Acid