Learn More
4-(Trifluoromethyl)benzoic acid, 98+%
CAS: 455-24-3 | C8H5F3O2 | 190.121 g/mol
$108.40 - $356.59
Chemical Identifiers
| CAS | 455-24-3 |
|---|---|
| Molecular Formula | C8H5F3O2 |
| Molecular Weight (g/mol) | 190.121 |
| MDL Number | MFCD00002562 |
| InChI Key | SWKPKONEIZGROQ-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride |
| PubChem CID | 9966 |
| ChEBI | CHEBI:60696 |
| IUPAC Name | 4-(trifluoromethyl)benzoic acid |
| SMILES | C1=CC(=CC=C1C(=O)O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1449406
|
Thermo Scientific Chemicals
A1449406 |
5 g |
Each for $108.40
|
|
|||||
|
AAA1449414
|
Thermo Scientific Chemicals
A1449414 |
25 g |
Each for $356.59
|
|
|||||
Description
4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N?-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N′-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 455-24-3 | |
| 190.121 | |
| SWKPKONEIZGROQ-UHFFFAOYSA-N | |
| 9966 | |
| 4-(trifluoromethyl)benzoic acid |
| C8H5F3O2 | |
| MFCD00002562 | |
| 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride | |
| CHEBI:60696 | |
| C1=CC(=CC=C1C(=O)O)C(F)(F)F |
Specifications
| 455-24-3 | |
| 247°C (753 mmHg) | |
| MFCD00002562 | |
| 2049241 | |
| Soluble in water. | |
| C1=CC(=CC=C1C(=O)O)C(F)(F)F | |
| 190.121 | |
| CHEBI:60696 | |
| ≥98% |
| 220°C to 222°C | |
| C8H5F3O2 | |
| 5 g | |
| 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride | |
| SWKPKONEIZGROQ-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzoic acid | |
| 9966 | |
| 190.12 | |
| 4-(Trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-242-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only