missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoic acid, 98+%

CAS: 455-24-3 | C8H5F3O2 | 190.121 g/mol

Supplier:  Thermo Scientific Chemicals A1449406

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1449406


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N?-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N′-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Spécifications

Spécifications

4-(Trifluoromethyl)benzoic acid
220°C to 222°C
C8H5F3O2
5 g
4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride
SWKPKONEIZGROQ-UHFFFAOYSA-N
4-(trifluoromethyl)benzoic acid
9966
190.12
455-24-3
247°C (753 mmHg)
MFCD00002562
2049241
Soluble in water.
C1=CC(=CC=C1C(=O)O)C(F)(F)F
190.121
CHEBI:60696
≥98%

Identifiants chimiques

455-24-3
190.121
SWKPKONEIZGROQ-UHFFFAOYSA-N
9966
4-(trifluoromethyl)benzoic acid
C8H5F3O2
MFCD00002562
4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride
CHEBI:60696
C1=CC(=CC=C1C(=O)O)C(F)(F)F
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 207-242-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only