missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoic acid, 98+%

Catalog No. AAA1449414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1449414 25 g
AAA1449406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1449414 Supplier Thermo Scientific Chemicals Supplier No. A1449414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 455-24-3 | C8H5F3O2 | 190.121 g/mol

4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N?-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N′-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 455-24-3
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00002562
InChI Key SWKPKONEIZGROQ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride
PubChem CID 9966
ChEBI CHEBI:60696
IUPAC Name 4-(trifluoromethyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)C(F)(F)F

Specifications

Melting Point 220°C to 222°C
Boiling Point 247°C (753 mmHg)
Quantity 25 g
Beilstein 2049241
Solubility Information Soluble in water.
Formula Weight 190.12
Percent Purity ≥98%
Chemical Name or Material 4-(Trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-242-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only