missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzhydrol, 97%

Catalog No. p-7046384
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1414703 1 g
AAL1414706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1414703 Supplier Thermo Scientific Chemicals Supplier No. L1414703

CAS: 395-23-3 | C14H11F3O | 252.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 395-23-3
Molecular Formula C14H11F3O
Molecular Weight (g/mol) 252.236
MDL Number MFCD00014399
InChI Key LIZDGCXCDJOWBS-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzhydrol, phenyl 4-trifluoromethyl phenyl methanol, benzhydrol, 4-trifluoromethyl-,, phenyl-4-trifluoromethyl phenyl methanol, phenyl 4-trifluoromethyl phenyl methan-1-ol, phenyl-4-trifluoromethyl-phenyl, acmc-20amtp, 4-trifluoromethylbenzhydrol, 4-trifluoromethylphenyl phenyl methanol
PubChem CID 258897
IUPAC Name phenyl-[4-(trifluoromethyl)phenyl]methanol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)C(F)(F)F)O

Specifications

Melting Point 59°C to 61°C
Boiling Point 122°C to 124°C (0.4 mmHg)
Quantity 1 g
Beilstein 2127184
Formula Weight 252.23
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzhydrol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-917-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only