missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzhydrazide, 98%

Catalog No. AAL0572403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0572403 1 g
AAL0572406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0572403 Supplier Thermo Scientific Chemicals Supplier No. L0572403

CAS: 339-59-3 | C8H7F3N2O | 204.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 339-59-3
Molecular Formula C8H7F3N2O
Molecular Weight (g/mol) 204.15
MDL Number MFCD00051703
InChI Key GKBDXTNCBPZMFX-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzhydrazide, 4-trifluoromethyl benzohydrazide, 4-trifluoromethyl benzoic acid hydrazide, 4-trifluoromethylbenzhydrazide, 4-trifluoromethylbenzoic acid hydrazide, 4-trifluoromethyl benzene-1-carbohydrazide, 1-trifluoromethyl benzene-4-carbohydrazide, alpha,alpha,alpha-trifluoro-p-toluic acid hydrazide, benzoic acid, 4-trifluoromethyl-, hydrazide, pubchem4506
PubChem CID 520662
IUPAC Name 4-(trifluoromethyl)benzohydrazide
SMILES NNC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 117°C to 121°C
Quantity 1 g
Beilstein 1968848
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only