missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzamidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T31151G
Specifications
| 4-(Trifluoromethyl)benzamidine Hydrochloride | |
| 167°C | |
| C8H8ClF3N2 | |
| 1 g | |
| C1=CC(=CC=C1C(=N)N)C(F)(F)F.Cl | |
| 224.611 | |
| 224.61 | |
| Crystalline Powder |
| 38980-96-0 | |
| White | |
| MFCD04114431 | |
| DKIFMADLURULQV-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzenecarboximidamide;hydrochloride | |
| 12535791 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 38980-96-0 | |
| 224.611 | |
| DKIFMADLURULQV-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzenecarboximidamide;hydrochloride |
| C8H8ClF3N2 | |
| MFCD04114431 | |
| 12535791 | |
| C1=CC(=CC=C1C(=N)N)C(F)(F)F.Cl |
Safety and Handling
TSCA : No