missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzamidine Hydrochloride 98.0+%, TCI America™

Catalog No. T3115200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3115200MG 200 mg
T31151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3115200MG Supplier TCI America Supplier No. T3115200MG
Only null left

Chemical Identifiers

CAS 38980-96-0
Molecular Formula C8H8ClF3N2
Molecular Weight (g/mol) 224.611
MDL Number MFCD04114431
InChI Key DKIFMADLURULQV-UHFFFAOYSA-N
PubChem CID 12535791
IUPAC Name 4-(trifluoromethyl)benzenecarboximidamide;hydrochloride
SMILES C1=CC(=CC=C1C(=N)N)C(F)(F)F.Cl

Specifications

Melting Point 167°C
Color White
Quantity 200 mg
Formula Weight 224.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)benzamidine Hydrochloride