missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzaldehyde 95.0+%, TCI America™
$72.70 - $833.24
Chemical Identifiers
| CAS | 455-19-6 |
|---|---|
| Molecular Formula | C8H5F3O |
| Molecular Weight (g/mol) | 174.122 |
| MDL Number | MFCD00006952 |
| InChI Key | BEOBZEOPTQQELP-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde |
| PubChem CID | 67996 |
| IUPAC Name | 4-(trifluoromethyl)benzaldehyde |
| SMILES | C1=CC(=CC=C1C=O)C(F)(F)F |
Chemical Identifiers
| 455-19-6 | |
| 174.122 | |
| BEOBZEOPTQQELP-UHFFFAOYSA-N | |
| 67996 | |
| C1=CC(=CC=C1C=O)C(F)(F)F |
| C8H5F3O | |
| MFCD00006952 | |
| 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde | |
| 4-(trifluoromethyl)benzaldehyde |
Specifications
| 455-19-6 | |
| 64°C | |
| MFCD00006952 | |
| 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde | |
| C1=CC(=CC=C1C=O)C(F)(F)F | |
| 174.122 | |
| 174.12 | |
| Liquid |
| Yellow | |
| C8H5F3O | |
| 5 g | |
| BEOBZEOPTQQELP-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzaldehyde | |
| 67996 | |
| ≥95.0% (GC) | |
| 4-(Trifluoromethyl)benzaldehyde |
Safety and Handling
EINECSNumber : (3)-3799
TSCA : Yes