Learn More
4-(Trifluoromethyl)benzaldehyde, 97%
CAS: 455-19-6 | C8H5F3O | 174.122 g/mol
$101.15 - $289.51
Chemical Identifiers
| CAS | 455-19-6 |
|---|---|
| Molecular Formula | C8H5F3O |
| Molecular Weight (g/mol) | 174.122 |
| MDL Number | MFCD00006952 |
| InChI Key | BEOBZEOPTQQELP-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde |
| PubChem CID | 67996 |
| IUPAC Name | 4-(trifluoromethyl)benzaldehyde |
| SMILES | C1=CC(=CC=C1C=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1527606
|
Thermo Scientific Chemicals
A1527606 |
5 g |
Each for $101.15
|
|
|||||
|
AAA1527614
|
Thermo Scientific Chemicals
A1527614 |
25 g |
Each for $289.51
|
|
|||||
Description
4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)benzaldehyde is useful reagent in kinetic studies of the asymmetric synthesis of alcohols and of the Wittig reaction. It is also used as Pharmaceutical intermediates.
Solubility
Soluble in water. 1.5 g/L at 20°C
Notes
Air Sensitive. Store away from air and oxidizing agents. Store under an inert atmosphere. Incompatible with oxidizing agents, reducing agents.
Chemical Identifiers
| 455-19-6 | |
| 174.122 | |
| BEOBZEOPTQQELP-UHFFFAOYSA-N | |
| 67996 | |
| C1=CC(=CC=C1C=O)C(F)(F)F |
| C8H5F3O | |
| MFCD00006952 | |
| 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde | |
| 4-(trifluoromethyl)benzaldehyde |
Specifications
| 455-19-6 | |
| 1.275 | |
| 65°C (149°F) | |
| 1.463 | |
| 5 g | |
| Air Sensitive | |
| Soluble in water. 1.5g/L at 20°C | |
| C1=CC(=CC=C1C=O)C(F)(F)F | |
| 174.122 | |
| 174.12 | |
| 4-(Trifluoromethyl)benzaldehyde |
| 1°C to 2°C | |
| 80°C to 81°C (25 mmHg) | |
| C8H5F3O | |
| MFCD00006952 | |
| 1101680 | |
| 4-trifluoromethyl benzaldehyde, benzaldehyde, 4-trifluoromethyl, p-trifluoromethylbenzaldehyde, p-trifluoromethyl benzaldehyde, alpha,alpha,alpha-trifluoro-p-tolualdehyde, 4'-trifluoromethyl benzaldehyde, p-trifluoromethyl-benzaldhyde, 4-trifluoromethyl-benzaldehyde | |
| BEOBZEOPTQQELP-UHFFFAOYSA-N | |
| 4-(trifluoromethyl)benzaldehyde | |
| 67996 | |
| 97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H320-H335
EINECSNumber : 207-240-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only