Learn More
4-(Trifluoromethyl)anisole, 98%
CAS: 402-52-8 | C8H7F3O | 176.138 g/mol
$45.53 - $421.90
Chemical Identifiers
| CAS | 402-52-8 |
|---|---|
| Molecular Formula | C8H7F3O |
| Molecular Weight (g/mol) | 176.138 |
| MDL Number | MFCD03094456 |
| InChI Key | CFIPQRIPCRRISV-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl anisole, 1-methoxy-4-trifluoromethyl benzene, 4-trifluoromethylanisole, benzene, 1-methoxy-4-trifluoromethyl, 4-methoxybenzotrifluoride, buttpark 180\07-83, p-trifluoromethylanisole, pubchem10126, 4-trifluoromethyl-anisol, 1-methoxy-4-trifluoromethyl-benzene |
| PubChem CID | 600604 |
| IUPAC Name | 1-methoxy-4-(trifluoromethyl)benzene |
| SMILES | COC1=CC=C(C=C1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH64187MD
|
Thermo Scientific Chemicals
H64187MD |
250 mg |
Each for $45.53
|
|
|||||
|
AAH6418703
|
Thermo Scientific Chemicals
H6418703 |
1 g |
Each for $111.38
|
|
|||||
|
AAH6418706
|
Thermo Scientific Chemicals
H6418706 |
5 g |
Each for $421.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 402-52-8 | |
| 176.138 | |
| CFIPQRIPCRRISV-UHFFFAOYSA-N | |
| 600604 | |
| COC1=CC=C(C=C1)C(F)(F)F |
| C8H7F3O | |
| MFCD03094456 | |
| 4-trifluoromethyl anisole, 1-methoxy-4-trifluoromethyl benzene, 4-trifluoromethylanisole, benzene, 1-methoxy-4-trifluoromethyl, 4-methoxybenzotrifluoride, buttpark 180\07-83, p-trifluoromethylanisole, pubchem10126, 4-trifluoromethyl-anisol, 1-methoxy-4-trifluoromethyl-benzene | |
| 1-methoxy-4-(trifluoromethyl)benzene |
Specifications
| 402-52-8 | |
| Colorless to Yellow | |
| 169°C (754 mmHg) | |
| C8H7F3O | |
| MFCD03094456 | |
| UN1993 | |
| CFIPQRIPCRRISV-UHFFFAOYSA-N | |
| 1-methoxy-4-(trifluoromethyl)benzene | |
| 600604 | |
| 98% | |
| 4-(Trifluoromethyl)anisole |
| -9°C | |
| 1.245 g/mL | |
| 60°C (140°F) | |
| 1.445 | |
| 250 mg | |
| 4-trifluoromethyl anisole, 1-methoxy-4-trifluoromethyl benzene, 4-trifluoromethylanisole, benzene, 1-methoxy-4-trifluoromethyl, 4-methoxybenzotrifluoride, buttpark 180\07-83, p-trifluoromethylanisole, pubchem10126, 4-trifluoromethyl-anisol, 1-methoxy-4-trifluoromethyl-benzene | |
| COC1=CC=C(C=C1)C(F)(F)F | |
| 176.138 | |
| 176.14 | |
| Liquid |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only