Learn More
4-Trifluoromethyl-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals
$301.86 - $301.86
Chemical Identifiers
| CAS | 163295-75-8 |
|---|---|
| Molecular Formula | C8H6ClF3O2S |
| Molecular Weight (g/mol) | 258.64 |
| MDL Number | MFCD01631932 |
| InChI Key | KKBNUPMMAGEQAT-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene |
| PubChem CID | 594332 |
| IUPAC Name | [4-(trifluoromethyl)phenyl]methanesulfonyl chloride |
| SMILES | FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2639503
|
Thermo Scientific Chemicals
H2639503 |
1 g |
Each for $301.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 163295-75-8 | |
| 258.64 | |
| KKBNUPMMAGEQAT-UHFFFAOYSA-N | |
| 594332 | |
| FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1 |
| C8H6ClF3O2S | |
| MFCD01631932 | |
| 4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene | |
| [4-(trifluoromethyl)phenyl]methanesulfonyl chloride |
Specifications
| 163295-75-8 | |
| C8H6ClF3O2S | |
| 1 g | |
| Moisture sensitive | |
| KKBNUPMMAGEQAT-UHFFFAOYSA-N | |
| [4-(trifluoromethyl)phenyl]methanesulfonyl chloride | |
| 594332 | |
| 97% |
| 103°C to 105°C | |
| MFCD01631932 | |
| UN3261 | |
| 4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene | |
| FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1 | |
| 258.64 | |
| 258.65 | |
| 4-Trifluoromethyl-alpha-toluenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only