missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. AAH2639503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2639503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2639503 Supplier Thermo Scientific Chemicals Supplier No. H2639503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 163295-75-8 | Molecular Formula: C8H6ClF3O2S | Molecular Weight: 258.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163295-75-8
Molecular Formula C8H6ClF3O2S
Molecular Weight (g/mol) 258.64
MDL Number MFCD01631932
InChI Key KKBNUPMMAGEQAT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene
PubChem CID 594332
IUPAC Name [4-(trifluoromethyl)phenyl]methanesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1

Specifications

Melting Point 103°C to 105°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 258.65
Percent Purity 97%
Chemical Name or Material 4-Trifluoromethyl-alpha-toluenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only