missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-(Trifluoromethyl)acetophenone 98.0+%, TCI America™
Supplier: TCI America T178425G
Specifications
| 4′-(Trifluoromethyl)acetophenone | |
| White-Yellow | |
| C9H7F3O | |
| 25 g | |
| HHAISVSEJFEWBZ-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)phenyl]ethanone | |
| 69731 | |
| ≥98.0% (GC) |
| 709-63-7 | |
| 80°C | |
| MFCD00000401 | |
| 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl | |
| CC(=O)C1=CC=C(C=C1)C(F)(F)F | |
| 188.149 | |
| 188.15 | |
| Crystalline Lumps |
Chemical Identifiers
| 709-63-7 | |
| 188.149 | |
| HHAISVSEJFEWBZ-UHFFFAOYSA-N | |
| 69731 | |
| CC(=O)C1=CC=C(C=C1)C(F)(F)F |
| C9H7F3O | |
| MFCD00000401 | |
| 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl | |
| 1-[4-(trifluoromethyl)phenyl]ethanone |
Safety and Handling
TSCA : No