missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-(Trifluoromethyl)acetophenone 98.0+%, TCI America™
$38.51 - $288.77
Chemical Identifiers
| CAS | 709-63-7 |
|---|---|
| Molecular Formula | C9H7F3O |
| Molecular Weight (g/mol) | 188.149 |
| MDL Number | MFCD00000401 |
| InChI Key | HHAISVSEJFEWBZ-UHFFFAOYSA-N |
| Synonym | 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl |
| PubChem CID | 69731 |
| IUPAC Name | 1-[4-(trifluoromethyl)phenyl]ethanone |
| SMILES | CC(=O)C1=CC=C(C=C1)C(F)(F)F |
Chemical Identifiers
| 709-63-7 | |
| 188.149 | |
| HHAISVSEJFEWBZ-UHFFFAOYSA-N | |
| 69731 | |
| CC(=O)C1=CC=C(C=C1)C(F)(F)F |
| C9H7F3O | |
| MFCD00000401 | |
| 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl | |
| 1-[4-(trifluoromethyl)phenyl]ethanone |
Specifications
| 709-63-7 | |
| 80°C | |
| MFCD00000401 | |
| 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl | |
| CC(=O)C1=CC=C(C=C1)C(F)(F)F | |
| 188.149 | |
| 188.15 | |
| Crystalline Lumps |
| White-Yellow | |
| C9H7F3O | |
| 1 g | |
| HHAISVSEJFEWBZ-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)phenyl]ethanone | |
| 69731 | |
| ≥98.0% (GC) | |
| 4′-(Trifluoromethyl)acetophenone |
Safety and Handling
TSCA : No