missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)acetanilide, 98+%

Catalog No. AAA1278914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1278914 25 g
AAA1278906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1278914 Supplier Thermo Scientific Chemicals Supplier No. A1278914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 349-97-3 | C9H8F3NO | 203.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 349-97-3
Molecular Formula C9H8F3NO
Molecular Weight (g/mol) 203.164
MDL Number MFCD00013562
InChI Key DFDHFECLWHHELH-UHFFFAOYSA-N
Synonym 4-trifluoromethyl acetanilide, n-4-trifluoromethyl phenyl acetamide, 4'-trifluoromethyl acetanilide, 4-trifluoromethylacetanilide, p-acetaminobenzotrifluoride, p-acetamidobenzotrifluoride, p-trifluoromethylacetanilide, p-acetylaminobenzotrifluoride, p-trifluoromethyl acetanilide
PubChem CID 67685
IUPAC Name N-[4-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 150°C to 154°C
Odor Odorless
Quantity 25 g
Beilstein 2106965
Formula Weight 203.16
Percent Purity ≥98%
Chemical Name or Material 4'-(Trifluoromethyl)acetanilide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 206-495-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only