missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-2-pyridone 98.0+%, TCI America™

Catalog No. T27901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27901G 1 g
T27905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27901G Supplier TCI America Supplier No. T27901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50650-59-4
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD00153195,MFCD09029339
InChI Key IKHLLNMSMFVTLP-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-2-ol, 4-trifluoromethyl pyridin-2 1h-one, 4-trifluoromethyl-pyridin-2-ol, 4-trifluoromethyl-2-pyridone, 2-hydroxy-4-trifluoromethylpyridine, 4-trifluoromethyl-2 1h pyridinone, 4-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 4-trifluoromethyl, 4-trifluoromethyl-2 1h-pyridinone
PubChem CID 2775130
IUPAC Name 4-(trifluoromethyl)-1,2-dihydropyridin-2-one
SMILES FC(F)(F)C1=CC(=O)NC=C1

Specifications

Melting Point 158°C
Quantity 1 g
Formula Weight 163.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)-2-pyridone