missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-2(1H)-quinolinone, 97%, Thermo Scientific Chemicals

Catalog No. AAH6173203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6173203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6173203 Supplier Thermo Scientific Chemicals Supplier No. H6173203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25199-84-2 | Molecular Formula: C10H6F3NO | Molecular Weight: 213.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25199-84-2
Molecular Formula C10H6F3NO
Molecular Weight (g/mol) 213.159
MDL Number MFCD03407380
InChI Key UUROBWTVZZNDFD-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl quinoline, 4-trifluoromethyl quinolin-2-ol, 4-trifluoromethyl-2 1h-quinolinone, 4-trifluoromethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-trifluoromethyl, 4-trifluoromethyl quinolin-2 1h-one, pubchem5867, ksc495g1t, 4-trifluoromethylquinolin-2-one, 4-trifluoromethyl-quinolin-2-ol
PubChem CID 2759347
IUPAC Name 4-(trifluoromethyl)-1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)C(F)(F)F

Specifications

Color White
Quantity 1 g
Formula Weight 213.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Trifluoromethyl-2(1H)-quinolinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only