missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals

Catalog No. p-7039762
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5916503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5916503 Supplier Thermo Scientific Chemicals Supplier No. H5916503

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68755-42-0
Molecular Formula C10H7F3O
Molecular Weight (g/mol) 200.16
MDL Number MFCD07772121
InChI Key LJVBFMQEZSEGRL-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1-indanone, 4-trifluoromethyl-2,3-dihydro-1h-inden-1-one, 4-trifluoromethyl-2,3-dihydroinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-trifluoromethyl, 4-trifluoromethyl indan-1-one, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one, 2,3-dihydro-1-oxo-4-trifluoromethyl-1h-indene, acmc-1b8x3, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one
PubChem CID 21921093
IUPAC Name 4-(trifluoromethyl)-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C(=CC=C2)C(F)(F)F

Specifications

Melting Point 38°C to 42°C
Boiling Point 67°C to 72°C (0.1 mmHg)
Flash Point >108°C (226°F)
Quantity 1 g
Formula Weight 200.16
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)-1-indanone
Hazard Category H302-H317
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only