accessibility menu, dialog, popup

UserName

OFFF

CAS RN 68755-42-0

IUPAC Name: 4-(trifluoromethyl)-2,3-dihydro-1H-inden-1-one
Synonyms: 4-(trifluoromethyl)-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 200.16
Molecular Formula: C10H7F3O
InChi Key: LJVBFMQEZSEGRL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC2=C1CCC2=O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results