missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)toluene 98.0+%, TCI America™

Supplier:  TCI America T24541G

 View more versions of this product

Catalog No. T24541G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(Trifluoromethoxy)toluene
Yellow
C8H7F3O
1 g
1-methyl-4-trifluoromethoxy benzene, 4-trifluoromethoxytoluene, 4-trifluoromethoxy toluene, 4-toluene trifluoromethyl ether, 4-methyltrifluoromethoxybenzene, benzene, 1-methyl-4-trifluoromethoxy, 4-methyl-alpha,alpha,alpha-trifluoroanisole, pubchem13486, acmc-209ofc
CC1=CC=C(C=C1)OC(F)(F)F
176.138
176.14
Liquid
706-27-4
135°C
MFCD00042413
1993
JUXFXYQUXNXVAA-UHFFFAOYSA-N
1-methyl-4-(trifluoromethoxy)benzene
2760631
≥98.0% (GC)

Chemical Identifiers

706-27-4
176.138
JUXFXYQUXNXVAA-UHFFFAOYSA-N
2760631
CC1=CC=C(C=C1)OC(F)(F)F
C8H7F3O
MFCD00042413
1-methyl-4-trifluoromethoxy benzene, 4-trifluoromethoxytoluene, 4-trifluoromethoxy toluene, 4-toluene trifluoromethyl ether, 4-methyltrifluoromethoxybenzene, benzene, 1-methyl-4-trifluoromethoxy, 4-methyl-alpha,alpha,alpha-trifluoroanisole, pubchem13486, acmc-209ofc
1-methyl-4-(trifluoromethoxy)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions