missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. T24151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24151G 1 g
T24155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24151G Supplier TCI America Supplier No. T24151G

Chemical Identifiers

CAS 133115-72-7
Molecular Formula C7H8ClF3N2O
Molecular Weight (g/mol) 228.60
MDL Number MFCD00053033
InChI Key KQXZVSQCMVKMBK-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenylhydrazine hydrochloride, 4-trifluoromethoxy phenyl hydrazine hydrochloride, 4-trifluoromethoxyphenylhydrazine hydrochloride, 4-trifluoromethoxy phenylhydrazine hcl, 4-trifluoromethoxyphenyl-hydrazine hcl, 1-4-trifluoromethoxy phenyl hydrazine hydrochloride, p-trifluoromethoxy phenylhydrazine hydrochloride, hydrazine, 4-trifluoromethoxy phenyl-, monohydrochloride
PubChem CID 2777328
IUPAC Name [4-(trifluoromethoxy)phenyl]hydrazine hydrochloride
SMILES Cl.NNC1=CC=C(OC(F)(F)F)C=C1

Specifications

Melting Point 230°C
Quantity 1 g
Formula Weight 228.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethoxy)phenylhydrazine Hydrochloride