missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylacetic acid, 98%

Catalog No. AAA1951206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1951206 5 g
AAA1951203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1951206 Supplier Thermo Scientific Chemicals Supplier No. A1951206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4315-07-5 | C9H6F3O3 | 219.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4315-07-5
MDL Number MFCD00061239

Specifications

Melting Point 86°C to 88°C
Molecular Formula C9H6F3O3
Quantity 5 g
Beilstein 2695336
Synonym 4-trifluoromethoxy phenylacetic acid, 2-4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenylaceticacid, 4-trifluoromethoxy-phenyl-acetic acid, 4-trifluoromethoxyphenylacetic acid, benzeneacetic acid, 4-trifluoromethoxy, maybridge3_006163
InChI Key ZMTIBXQPXBUWPQ-UHFFFAOYSA-M
SMILES C1=CC(=CC=C1CC(=O)O)OC(F)(F)F
IUPAC Name 2-[4-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol) 219.14
PubChem CID 2777319
Formula Weight 220.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only